Assay Details
| Gene Symbol: LOC103689951 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006230629.2 | XP_006230691.1 | 1 - 2 | 273 | 100 | |
| Gene Symbol: Ppfia1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106320.2 | NP_001099790.2 | 20 - 21 | 2864 | 100 | |
| XM_006230721.2 | XP_006230783.1 | 22 - 23 | 2970 | 100 | ||
| XM_006230722.2 | XP_006230784.1 | 20 - 21 | 2865 | 100 | ||
| XM_006230723.2 | XP_006230785.1 | 20 - 21 | 2865 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

LOC103689951
Genomic Map