Assay Details
| Gene Symbol: LOC100912380 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006228796.2 | XP_006228858.2 | 4 - 5 | 228 | 78 | |
| Gene Symbol: Capns1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017118.1 | NP_058814.1 | 5 - 6 | 452 | 78 | |
| GenBank mRNA | BC098068.1 | - | 5 - 6 | 452 | 7462537 | 78 |
| FQ229527.1 | - | 135 | 78 | |||
| U10861.1 | - | 2 - 3 | 141 | 78 | ||
| U53859.1 | - | 4 - 5 | 388 | 78 | ||
| Gene Symbol: LOC100911363 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003748836.3 | XP_003748884.1 | 5 - 6 | 501 | 78 | |
| XM_008759263.1 | XP_008757485.1 | 5 - 6 | 703 | 78 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100912380
Genomic Map