Assay Details
| Gene Symbol: Ppm1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006240193.2 | XP_006240255.1 | 6 | 10699 | 153 | |
| XM_006240194.2 | XP_006240256.1 | 6 | 7653 | 153 | ||
| XM_006240195.2 | XP_006240257.1 | 6 | 7417 | 153 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ppm1a
Genomic Map