Assay Details
| Gene Symbol: Baz1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170568.1 | NP_001164039.1 | 8 - 9 | 1253 | 69 | |
| GenBank mRNA | CB614294.1 | - | 4 - 5 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Baz1a
Genomic Map