Assay Details
Gene Symbol: Rdh11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012193.1 | NP_001012193.1 | 4 - 5 | 530 | 83 | |
GenBank mRNA | AY387097.1 | - | 4 - 5 | 698 | 83 | |
BC079276.1 | - | 4 - 5 | 530 | 7134077 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
