Assay Details
Gene Symbol: Cldn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031700.2 | NP_113888.2 | 1 | 1363 | 81 | |
GenBank mRNA | BC062411.1 | - | 1 | 988 | 5623891 | 81 |
M74067.1 | - | 1 | 1082 | 81 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
