Assay Details
| Gene Symbol: Sdsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108336.1 | NP_001101806.1 | 4 - 5 | 615 | 107 | |
| XM_006249399.2 | XP_006249461.1 | 2 - 3 | 596 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sdsl
Genomic Map