Assay Details
Gene Symbol: Cdk2ap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769238.1 | XP_008767460.1 | 1 - 2 | 614 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008769238.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
