Assay Details
| Gene Symbol: Asphd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008769345.1 | XP_008767567.1 | 2 - 3 | 1485 | 126 | |
| XM_008769347.1 | XP_008767569.1 | 3 - 4 | 675 | 126 | ||
| XM_008769348.1 | XP_008767570.1 | 3 - 4 | 1388 | 126 | ||
| XM_008769349.1 | XP_008767571.1 | 3 - 4 | 1271 | 126 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008769345.1, XM_008769347.1, XM_008769348.1, XM_008769349.1 transcripts.
Set Membership: |
|

Asphd2
Genomic Map