Assay Details
| Gene Symbol: Arl6ip4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025630.1 | NP_001020801.1 | 3 - 4 | 906 | 92 | |
| GenBank mRNA | BC097374.1 | - | 3 - 4 | 906 | 7444619 | 92 |
More Information
Updated Mapping Information
Set Membership: |
|

Arl6ip4
Genomic Map