Assay Details
Gene Symbol: Ran | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006249285.2 | XP_006249347.1 | 1 - 2 | 54 | 80 | |
GenBank mRNA | BC059123.1 | - | 1 - 2 | 82 | 6920235 | 80 |
FQ220083.1 | - | 1 - 2 | 50 | 80 | ||
FQ220326.1 | - | 1 - 2 | 50 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC059123.1,XM_006249285.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
