Assay Details
| Gene Symbol: Chfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009258.1 | NP_001009258.1 | 16 - 17 | 1928 | 64 | |
| XM_006249535.2 | XP_006249597.1 | 16 - 17 | 2010 | 64 | ||
| XM_006249536.2 | XP_006249598.1 | 15 - 16 | 1890 | 64 | ||
| GenBank mRNA | BC087162.1 | - | 16 - 17 | 1928 | 7135077 | 64 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
mitotic cell cycle
ubiquitin-dependent protein catabolic process
mitotic cell cycle checkpoint
protein ubiquitination
modification-dependent protein catabolic process
positive regulation of protein ubiquitination
protein destabilization
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
nucleotide binding
ubiquitin-protein transferase activity
zinc ion binding
ubiquitin protein ligase activity
protein polyubiquitination
mitotic cell cycle
ubiquitin-dependent protein catabolic process
mitotic cell cycle checkpoint
protein ubiquitination
modification-dependent protein catabolic process
positive regulation of protein ubiquitination
protein destabilization
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
nucleotide binding
ubiquitin-protein transferase activity
zinc ion binding
ubiquitin protein ligase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P023689 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540372 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828339 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168178 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318759 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778866 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588463 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859982 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365499 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424852 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569933 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795572 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547106 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796872 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456400 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885195 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646337 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86414 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334974 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83214 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485193 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54423 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140476 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162958 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57879 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P023689 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P023689 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P023689 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P449969 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1046651 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 6563 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 42193 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 11517 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10763123 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763118 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763129 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763120 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763122 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763128 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763117 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763124 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763121 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763130 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763133 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763131 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763134 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763125 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763126 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763127 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763132 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763119 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10763116 | - | - |
| Rat Genome 230 2.0 Array | 1383313_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1383313_at | - | - |
| Rat Genome 230 2.0 Array | 1393293_at | - | - |
| Rat Genome 230 2.0 Array | 1393293_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1393293_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1393293_at | - | - |
| Affymetrix HT Rat Focus Array | 1383313_at | - | - |
| Affymetrix HT Rat Focus Array | 1383313_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1200015991.rn.1 | TC1200001875.rn.1 | RTA TC |
| Rat Expression Set 230 | 1393293_PM_at | - | - |
| Rat Expression Set 230 | 1383313_PM_at | - | - |
| Rat Expression Set 230 | 1383313_at | - | - |
| Rat Expression Set 230 | 1393293_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763116 | - | - |
| Rat Genome U34 Set | rc_AA925391_at | - | - |
| Rat Genome U34 Set | rc_AI029628_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20788923 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1364233 | - | - |
| RatRef-12 Expression BeadChip | ILMN_57717 | - | - |

Chfr
Genomic Map