Assay Details
Gene Symbol: Sart3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107156.1 | NP_001100626.1 | 14 - 15 | 1760 | 62 | |
XM_006249501.2 | XP_006249563.1 | 14 - 15 | 2133 | 62 | ||
XM_006249502.2 | XP_006249564.1 | 15 - 16 | 1765 | 62 | ||
XM_008769338.1 | XP_008767560.1 | 14 - 15 | 2573 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
