Assay Details
Gene Symbol: Slc12a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134405.1 | NP_599232.1 | 11 | 1805 | 58 | |
GenBank mRNA | AB023645.1 | - | 11 | 1805 | 58 | |
BC081728.1 | - | 11 | 1838 | 7129648 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
