Assay Details
Gene Symbol: Hip1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134763.1 | NP_001128235.1 | 9 - 10 | 810 | 60 | |
NM_031234.1 | NP_112513.1 | 9 - 10 | 810 | 60 | ||
XM_008769266.1 | XP_008767488.1 | 9 - 10 | 899 | 60 | ||
XM_008769267.1 | XP_008767489.1 | 9 - 10 | 899 | 60 | ||
XM_008769268.1 | XP_008767490.1 | 9 - 10 | 2949 | 60 | ||
GenBank mRNA | AB005052.1 | - | 9 - 10 | 810 | 60 | |
BC169096.1 | - | 9 - 10 | 797 | 7936362 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
