Assay Details
| Gene Symbol: Rabgef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108333.1 | NP_001101803.1 | 4 - 5 | 835 | 80 | |
| XM_006249238.2 | XP_006249300.1 | 5 - 6 | 930 | 80 | ||
| XM_006249239.2 | XP_006249301.1 | 5 - 6 | 682 | 80 | ||
| XM_006249240.2 | XP_006249302.1 | 5 - 6 | 919 | 80 | ||
| XM_008769150.1 | XP_008767372.1 | 5 - 6 | 926 | 80 | ||
| GenBank mRNA | BC168697.1 | - | 2 - 3 | 234 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rabgef1
Genomic Map