Assay Details
| Gene Symbol: Rimbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100488.2 | NP_001093958.1 | 14 - 15 | 2896 | 78 | |
| XM_006249257.2 | XP_006249319.1 | 13 - 14 | 2858 | 78 | ||
| XM_006249258.2 | XP_006249320.1 | 13 - 14 | 3041 | 78 | ||
| XM_008769189.1 | XP_008767411.1 | 13 - 14 | 2790 | 78 | ||
| XM_008769190.1 | XP_008767412.1 | 13 - 14 | 3958 | 78 | ||
| XM_008769191.1 | XP_008767413.1 | 13 - 14 | 2858 | 78 | ||
| XM_008769192.1 | XP_008767414.1 | 13 - 14 | 2858 | 78 | ||
| XM_008769193.1 | XP_008767415.1 | 13 - 14 | 2858 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rimbp2
Genomic Map