Assay Details
| Gene Symbol: Fam216a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008290.1 | NP_001008291.1 | 2 - 3 | 387 | 93 | |
| GenBank mRNA | BC086384.1 | - | 2 - 3 | 387 | 7301624 | 93 |
| FQ220218.1 | - | 2 - 3 | 198 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam216a
Genomic Map