Assay Details
| Gene Symbol: Vps37d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128193.2 | NP_001121665.1 | 2 - 3 | 349 | 71 | |
| GenBank mRNA | BC166560.1 | - | 2 - 3 | 349 | 7314601 | 71 |
More Information
Updated Mapping Information
Set Membership: |
|

Vps37d
Genomic Map