Assay Details
| Gene Symbol: ST7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004102.1 | NP_001004102.1 | 14 - 15 | 1745 | 96 | |
| XM_006236132.2 | XP_006236194.1 | 15 - 16 | 2654 | 96 | ||
| XM_006236133.2 | XP_006236195.1 | 15 - 16 | 2947 | 96 | ||
| XM_006236134.2 | XP_006236196.1 | 15 - 16 | 2183 | 96 | ||
| XM_006236135.2 | XP_006236197.1 | 14 - 15 | 2114 | 96 | ||
| XM_008762700.1 | XP_008760922.1 | 15 - 16 | 3379 | 96 | ||
| XM_008762701.1 | XP_008760923.1 | 16 - 17 | 2341 | 96 | ||
| GenBank mRNA | BC078945.1 | - | 16 - 17 | 2001 | 96 | |
| BC099820.1 | - | 13 - 14 | 1734 | 7930680 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P008928 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P160418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972872 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264698 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509141 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167157 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12770 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876772 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369523 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184132 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868296 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203229 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292296 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276946 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418297 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316837 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815414 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572412 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234373 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592420 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22221 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210768 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152937 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604391 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964240 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617747 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623570 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945973 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282987 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941942 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689857 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305170 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654072 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373254 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952817 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848271 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698812 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574226 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30830 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361696 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58950 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726103 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136266 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58940 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706598 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815150 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586643 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667652 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901361 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163409 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930849 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162485 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725307 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497522 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398578 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106564 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305932 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554337 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940499 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206440 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177813 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933673 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446517 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951726 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614952 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623261 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149910 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690061 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363249 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345800 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30655 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313346 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232086 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706333 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468225 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262619 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55141 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846778 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906726 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468617 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649004 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203840 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465106 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125560 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733780 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408374 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338068 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192019 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443279 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818703 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27004 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278286 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709640 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949674 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263761 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837106 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917627 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296398 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313351 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893075 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19845 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510088 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62130 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849832 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356542 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517374 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165859 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50877 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534806 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41866 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240601 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527207 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467484 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889941 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71711 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351662 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664595 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715887 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697701 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85142 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112894 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329554 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769854 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883939 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129334 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240414 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585385 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477885 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943743 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265574 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930845 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2335 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926269 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845719 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615330 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76337 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72255 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9046 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23644 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77409 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115455 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55761 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144652 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141942 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20648 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143642 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138659 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110587 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92447 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33091 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178892 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58171 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35655 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39730 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91124 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154152 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50705 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P160418 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P008928 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P160418 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P008928 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P160418 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P008928 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P266920 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 18889 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 81397 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 20343 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 76592 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 19724 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 35764 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 20116 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 59173 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 87534 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 75449 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 57302 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 49608 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 30328 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 76152 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 79056 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 62405 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 82851 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10853855 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853866 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853869 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853870 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853865 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853863 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853864 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853868 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853861 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853857 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853854 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853856 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853859 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853862 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853858 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853867 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10853852 | - | - |
| Rat Genome 230 2.0 Array | 1392925_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1392925_at | - | - |
| Affymetrix HT Rat Focus Array | 1392925_at | - | - |
| Affymetrix HT Rat Focus Array | 1392925_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0400003148.rn.1 | TC0400000436.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400003150.rn.1 | TC0400000436.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400003146.rn.1 | TC0400000436.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400003145.rn.1 | TC0400000436.rn.1 | RTA TC |
| Rat Expression Set 230 | 1392925_PM_at | - | - |
| Rat Expression Set 230 | 1392925_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853852 | - | - |
| Rat Genome U34 Set | rc_AI229362_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21343223 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376390 | - | - |

ST7
Genomic Map