Assay Details
Gene Symbol: LOC500077 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024325.1 | NP_001019496.1 | 2 - 3 | 553 | 84 | |
GenBank mRNA | BC079043.1 | - | 2 - 3 | 553 | 7114933 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
