Assay Details
| Gene Symbol: Fezf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109224.1 | NP_001102694.1 | 1 - 2 | 1091 | 107 | |
| XM_008762805.1 | XP_008761027.1 | 2 - 3 | 653 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fezf1
Genomic Map