Assay Details
| Gene Symbol: Fezf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109224.1 | NP_001102694.1 | 3 - 4 | 1353 | 119 | |
| XM_008762805.1 | XP_008761027.1 | 4 - 5 | 915 | 119 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fezf1
Genomic Map