Assay Details
Gene Symbol: Adck2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107855.1 | NP_001101325.1 | 3 - 4 | 1591 | 156 | |
XM_006236325.2 | XP_006236387.1 | 2 - 3 | 1468 | 156 | ||
XM_006236326.2 | XP_006236388.1 | 2 - 3 | 1467 | 156 | ||
XM_008762782.1 | XP_008761004.1 | 3 - 4 | 1614 | 156 | ||
GenBank mRNA | BC161918.1 | - | 2 - 3 | 1379 | 8369971 | 156 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
