Assay Details
| Gene Symbol: Hyal5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024321.1 | NP_001019492.1 | 1 - 2 | 106 | 116 | |
| GenBank mRNA | BC091219.1 | - | 1 - 2 | 106 | 7112082 | 116 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC091219.1, NM_001024321.1 transcripts.
Set Membership: |
|

Hyal5
Genomic Map