Assay Details
| Gene Symbol: Ndufb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108624.2 | NP_001102094.2 | 1 - 2 | 130 | 87 | |
| GenBank mRNA | BC166901.1 | - | 1 - 2 | 107 | 6922162 | 87 |
| C06706.1 | - | 1 - 2 | 130 | 87 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ndufb2
Genomic Map