Assay Details
Gene Symbol: Tpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134994.1 | NP_001128466.1 | 6 - 7 | 514 | 75 | |
XM_008762921.1 | XP_008761143.1 | 3 - 4 | 2060 | 75 | ||
GenBank mRNA | BC168695.1 | - | 6 - 7 | 403 | 7938123 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
