Assay Details
Gene Symbol: Cfl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017147.2 | NP_058843.1 | 2 - 3 | 396 | 77 | |
GenBank mRNA | BC059143.1 | - | 2 - 3 | 396 | 6889053 | 77 |
BC086533.1 | - | 2 - 3 | 381 | 7305422 | 77 | |
FQ212136.1 | - | 1 - 2 | 268 | 77 | ||
FQ212794.1 | - | 2 - 3 | 395 | 77 | ||
FQ212910.1 | - | 2 - 3 | 391 | 77 | ||
FQ213701.1 | - | 2 - 3 | 77 | |||
FQ213897.1 | - | 2 - 3 | 420 | 77 | ||
FQ214120.1 | - | 2 - 3 | 392 | 77 | ||
FQ214438.1 | - | 2 - 3 | 391 | 77 | ||
FQ220384.1 | - | 2 - 3 | 407 | 77 | ||
FQ220571.1 | - | 2 - 3 | 393 | 77 | ||
FQ226405.1 | - | 2 - 3 | 391 | 77 | ||
FQ227487.1 | - | 2 - 3 | 393 | 77 | ||
FQ229073.1 | - | 2 - 3 | 420 | 77 | ||
FQ229766.1 | - | 2 - 3 | 395 | 77 | ||
FQ229833.1 | - | 2 - 3 | 391 | 77 | ||
FQ229875.1 | - | 2 - 3 | 395 | 77 | ||
FQ230001.1 | - | 2 - 3 | 395 | 77 | ||
FQ230091.1 | - | 2 - 3 | 392 | 77 | ||
FQ231379.1 | - | 2 - 3 | 391 | 77 | ||
FQ232577.1 | - | 1 - 2 | 271 | 77 | ||
FQ233750.1 | - | 2 - 3 | 391 | 77 | ||
FQ234162.1 | - | 2 - 3 | 395 | 77 | ||
FQ234303.1 | - | 2 - 3 | 391 | 77 | ||
X62908.1 | - | 1 - 2 | 320 | 77 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
