Assay Details
| Gene Symbol: Dkk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106350.1 | NP_001099820.1 | 384 | 138 | ||
| GenBank mRNA | JN966752.1 | - | 416 | 138 | ||
| Gene Symbol: LOC100912050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749089.3 | XP_003749137.1 | 1 - 2 | 571 | 138 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dkk1
Genomic Map