Assay Details
| Gene Symbol: Dkk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106350.1 | NP_001099820.1 | 552 | 60 | ||
| GenBank mRNA | JN966752.1 | - | 584 | 60 | ||
| Gene Symbol: LOC100912050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749089.3 | XP_003749137.1 | 2 - 3 | 739 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dkk1
Genomic Map