Assay Details
| Gene Symbol: Plgrkt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106347.1 | NP_001099817.1 | 3 - 4 | 380 | 142 | |
| XM_006231205.2 | XP_006231267.1 | 3 - 4 | 284 | 142 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001106347.1, XM_006231205.2 transcripts.
Set Membership: |
|

Plgrkt
Genomic Map