Assay Details
| Gene Symbol: Plgrkt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106347.1 | NP_001099817.1 | 3 - 4 | 380 | 80 | |
| XM_006231203.2 | XP_006231265.1 | 4 - 5 | 441 | 80 | ||
| XM_006231204.2 | XP_006231266.1 | 4 - 5 | 345 | 80 | ||
| XM_006231205.2 | XP_006231267.1 | 3 - 4 | 284 | 80 | ||
| XM_008760323.1 | XP_008758545.1 | 1 - 2 | 470 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plgrkt
Genomic Map