Assay Details
| Gene Symbol: Gpr26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138841.1 | NP_620196.1 | 2 - 3 | 808 | 65 | |
| GenBank mRNA | AF208288.1 | - | 2 - 3 | 808 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpr26
Genomic Map