Assay Details
Gene Symbol: Tcirg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199089.2 | NP_954520.2 | 7 - 8 | 811 | 54 | |
XM_006230727.2 | XP_006230789.1 | 8 - 9 | 915 | 54 | ||
XM_006230728.2 | XP_006230790.1 | 8 - 9 | 886 | 54 | ||
XM_008760097.1 | XP_008758319.1 | 8 - 9 | 982 | 54 | ||
GenBank mRNA | BC061859.1 | - | 8 - 9 | 884 | 5598499 | 54 |
DQ286426.1 | - | 7 - 8 | 811 | 54 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
