Assay Details
| Gene Symbol: Cdkn1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033757.1 | NP_001028929.1 | 1 - 2 | 1214 | 75 | |
| XM_008760076.1 | XP_008758298.1 | 2 - 3 | 1126 | 75 | ||
| XM_008760077.1 | XP_008758299.1 | 2 - 3 | 1132 | 75 | ||
| XM_008760078.1 | XP_008758300.1 | 2 - 3 | 1086 | 75 | ||
| XM_008760079.1 | XP_008758301.1 | 2 - 3 | 1084 | 75 | ||
| GenBank mRNA | AY351678.1 | - | 1 - 2 | 1215 | 75 | |
| CF110214.1 | - | 1 - 2 | 359 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdkn1c
Genomic Map