Assay Details
Gene Symbol: Nap1l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012170.1 | NP_001012170.1 | 7 - 8 | 644 | 68 | |
XM_006230823.2 | XP_006230885.1 | 7 - 8 | 650 | 68 | ||
XM_006230824.2 | XP_006230886.1 | 8 - 9 | 717 | 68 | ||
XM_006230826.2 | XP_006230888.1 | 7 - 8 | 650 | 68 | ||
GenBank mRNA | BC085801.1 | - | 7 - 8 | 644 | 7110617 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
