Assay Details
Gene Symbol: Cyp2c11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019184.2 | NP_062057.2 | 1 - 2 | 195 | 99 | |
GenBank mRNA | BC088146.1 | - | 1 - 2 | 185 | 7369647 | 99 |
J02657.1 | - | 1 - 2 | 195 | 99 | ||
U33173.1 | - | 1 - 2 | 195 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
