Assay Details
Gene Symbol: Cyp2c11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019184.2 | NP_062057.2 | 6 - 7 | 983 | 75 | |
XM_008760364.1 | XP_008758586.1 | 5 - 6 | 811 | 75 | ||
GenBank mRNA | BC088146.1 | - | 6 - 7 | 973 | 7369647 | 75 |
J02657.1 | - | 6 - 7 | 983 | 75 | ||
U33173.1 | - | 6 - 7 | 983 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
