Assay Details
Gene Symbol: Phemx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002728846.4 | XP_002728892.3 | 7 - 8 | 3038 | 120 | |
XM_006230915.2 | XP_006230977.1 | 6 - 7 | 2963 | 120 | ||
XM_006230916.2 | XP_006230978.1 | 6 - 7 | 2951 | 120 | ||
GenBank mRNA | BC088266.1 | - | 4 - 5 | 329 | 120 | |
BC158837.1 | - | 5 - 6 | 503 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
