Assay Details
| Gene Symbol: Smpd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002729839.4 | XP_002729885.2 | 1 - 2 | 967 | 59 | |
| XM_006241911.2 | XP_006241973.1 | 1 - 2 | 1065 | 59 | ||
| GenBank mRNA | BC166812.1 | - | 1 - 2 | 979 | 59 | |
More Information
Set Membership: |
|

Smpd5
Genomic Map