Assay Details
| Gene Symbol: Smpd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002729839.4 | XP_002729885.2 | 5 - 6 | 1328 | 62 | |
| XM_006241911.2 | XP_006241973.1 | 4 - 5 | 1495 | 62 | ||
| XM_008765630.1 | XP_008763852.1 | 5 - 6 | 1340 | 62 | ||
| XM_008765631.1 | XP_008763853.1 | 4 - 5 | 1453 | 62 | ||
| GenBank mRNA | BC166812.1 | - | 4 - 5 | 1409 | 62 | |
| CB578696.1 | - | 3 - 4 | 401 | 62 | ||
| CB715946.1 | - | 3 - 4 | 382 | 62 | ||
More Information
Set Membership: |
|

Smpd5
Genomic Map