Assay Details
| Gene Symbol: Smpd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002729839.4 | XP_002729885.2 | 6 - 7 | 1471 | 150 | |
| XM_006241911.2 | XP_006241973.1 | 5 - 6 | 1638 | 150 | ||
| XM_008765630.1 | XP_008763852.1 | 6 - 7 | 1483 | 150 | ||
| XM_008765631.1 | XP_008763853.1 | 5 - 6 | 1596 | 150 | ||
| GenBank mRNA | BC166812.1 | - | 5 - 6 | 1552 | 150 | |
| CB578696.1 | - | 4 - 5 | 544 | 150 | ||
More Information
Set Membership: |
|

Smpd5
Genomic Map