Assay Details
Gene Symbol: Pmm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008323.1 | NP_001008324.1 | 4 - 5 | 411 | 76 | |
GenBank mRNA | BC086346.1 | - | 4 - 5 | 411 | 7302298 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
