Assay Details
Gene Symbol: Ccdc53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106776.1 | NP_001100246.1 | 2 - 3 | 301 | 109 | |
XM_006241199.2 | XP_006241261.1 | 2 - 3 | 273 | 109 | ||
GenBank mRNA | FQ221863.1 | - | 1 - 2 | 76 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
