Assay Details
| Gene Symbol: Ccdc53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106776.1 | NP_001100246.1 | 5 - 6 | 583 | 88 | |
| XM_006241199.2 | XP_006241261.1 | 5 - 6 | 552 | 88 | ||
| GenBank mRNA | FQ221863.1 | - | 4 - 5 | 358 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc53
Genomic Map