Assay Details
Gene Symbol: Ccdc53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106776.1 | NP_001100246.1 | 6 - 7 | 646 | 146 | |
XM_006241199.2 | XP_006241261.1 | 6 - 7 | 615 | 146 | ||
GenBank mRNA | FQ221863.1 | - | 5 - 6 | 421 | 146 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P600796 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119937 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565030 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160226 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626871 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254354 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739436 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642594 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766471 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646141 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276885 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492554 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840582 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745828 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729706 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535970 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828257 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702934 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144093 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22597 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242102 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390803 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295732 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185847 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176658 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815862 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633643 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952812 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669408 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115396 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49046 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96029 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71013 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P600796 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P600796 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P600796 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P600796 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 70429 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10894726 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894727 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894724 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894728 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894723 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894725 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894729 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10894722 | - | - |
Rat Genome 230 2.0 Array | 1375634_at | - | - |
Rat Genome 230 2.0 Array | 1375634_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1375634_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1375634_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0700003920.rn.1 | TC0700000451.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700003921.rn.1 | TC0700000451.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700003924.rn.1 | TC0700000451.rn.1 | RTA TC |
Rat Expression Set 230 | 1375634_at | - | - |
Rat Expression Set 230 | 1375634_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894722 | - | - |
Rat Genome U34 Set | rc_AA799472_at | - | - |
Rat Genome U34 Set | rc_AA799472_g_at | - | - |
Rat Genome U34 Set | rc_AI233259_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21238260 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1361976 | - | - |
