Assay Details
Gene Symbol: Ube2n | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053928.2 | NP_446380.1 | 3 - 4 | 755 | 106 | |
GenBank mRNA | BC090072.1 | - | 3 - 4 | 755 | 7113593 | 106 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
double-strand break repair via homologous recombination
DNA double-strand break processing
postreplication repair
ubiquitin-dependent protein catabolic process
protein ubiquitination
histone ubiquitination
positive regulation of histone modification
regulation of histone ubiquitination
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of DNA repair
T cell receptor signaling pathway
positive regulation of NF-kappaB transcription factor activity
positive regulation of ubiquitin-protein transferase activity
protein K63-linked ubiquitination
ubiquitin-protein transferase activity
ATP binding
ligase activity
ubiquitin-like protein transferase activity
ubiquitin protein ligase binding
ubiquitin binding
poly(A) RNA binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity
double-strand break repair via homologous recombination
DNA double-strand break processing
postreplication repair
ubiquitin-dependent protein catabolic process
protein ubiquitination
histone ubiquitination
positive regulation of histone modification
regulation of histone ubiquitination
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of DNA repair
T cell receptor signaling pathway
positive regulation of NF-kappaB transcription factor activity
positive regulation of ubiquitin-protein transferase activity
protein K63-linked ubiquitination
ubiquitin-protein transferase activity
ATP binding
ligase activity
ubiquitin-like protein transferase activity
ubiquitin protein ligase binding
ubiquitin binding
poly(A) RNA binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P042007 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839821 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851347 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831129 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698032 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367736 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277841 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228220 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672424 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405112 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339995 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584785 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560159 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172562 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159715 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72120 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420299 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477255 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319595 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503104 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52465 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164636 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852926 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250803 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165373 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 103384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39353 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53117 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P042007 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P042007 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P042007 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P10034 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P278439 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 46387 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 62921 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 77175 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10895030 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10720521 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10895033 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10895031 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10895032 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10895028 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10895029 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10759434 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10860547 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10759433 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10895027 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10720520 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10860546 | - | - |
Rat Genome 230 2.0 Array | 1369617_at | - | - |
Rat Genome 230 2.0 Array | 1369617_PM_at | - | - |
Rat Genome 230 2.0 Array | 1399143_PM_at | - | - |
Rat Genome 230 2.0 Array | 1399143_at | - | - |
Affymetrix HT Rat Focus Array | 1369617_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1399143_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1369617_at | - | - |
Affymetrix HT Rat Focus Array | 1399143_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0700004546.rn.1 | TC0700000546.rn.1 | RTA TC |
Rat Expression Set 230 | 1369617_at | - | - |
Rat Expression Set 230 | 1399143_PM_at | - | - |
Rat Expression Set 230 | 1369617_PM_at | - | - |
Rat Expression Set 230 | 1399143_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10860546 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10895027 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10759433 | - | - |
Rat Genome U34 Set | rc_AA925340_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21207604 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1651183 | - | - |
RatRef-12 Expression BeadChip | ILMN_52862 | - | - |
