Assay Details
Gene Symbol: Ccdc38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241271.2 | XP_006241333.1 | 15 - 16 | 1599 | 126 | |
XM_008765310.1 | XP_008763532.1 | 14 - 15 | 1329 | 126 | ||
XM_008765311.1 | XP_008763533.1 | 15 - 16 | 1611 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
