Assay Details
Gene Symbol: Rassf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108747.1 | NP_001102217.1 | 4 - 5 | 693 | 126 | |
XM_006241402.2 | XP_006241464.1 | 4 - 5 | 729 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P425848 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P132957 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225013 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274217 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756161 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90985 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448958 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745644 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7570 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803255 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659436 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31830 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598344 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293407 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800487 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321533 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342201 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 551608 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827016 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628345 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703468 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49333 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559614 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901707 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164237 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302176 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185621 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47667 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821165 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115094 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509586 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252933 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788229 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533807 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759229 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651705 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394336 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677963 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782383 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174047 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531526 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79739 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202319 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784028 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260029 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930527 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927722 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125310 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131668 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20974 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71015 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6691 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52381 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33523 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P425848 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P132957 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P132957 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P425848 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P425848 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P132957 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P551054 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 81996 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10902712 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902709 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902711 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902710 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902713 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10902708 | - | - |
Rat Genome 230 2.0 Array | 1373934_at | - | - |
Rat Genome 230 2.0 Array | 1373934_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1373934_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1373934_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0700021863.rn.1 | TC0700002821.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700021862.rn.1 | TC0700002821.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700021861.rn.1 | TC0700002821.rn.1 | RTA TC |
Rat Expression Set 230 | 1373934_at | - | - |
Rat Expression Set 230 | 1373934_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902708 | - | - |
Rat Genome U34 Set | rc_AA956928_at | - | - |
Rat Genome U34 Set | rc_AI178214_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21317433 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1358927 | - | - |
