Assay Details
| Gene Symbol: Anks1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271371.1 | NP_001258300.1 | 16 - 17 | 2620 | 68 | |
| XM_006241248.2 | XP_006241310.1 | 2 - 3 | 666 | 68 | ||
| XM_006241249.2 | XP_006241311.1 | 2 - 3 | 665 | 68 | ||
| XM_008765239.1 | XP_008763461.1 | 4 - 5 | 1001 | 68 | ||
| XM_008765240.1 | XP_008763462.1 | 2 - 3 | 666 | 68 | ||
| XM_008765241.1 | XP_008763463.1 | 4 - 5 | 1002 | 68 | ||
| XM_008765243.1 | XP_008763465.1 | 2 - 3 | 666 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Anks1b
Genomic Map